Assay Details
| Gene Symbol: LOC4328083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771656.1 | XP_015627142.1 | 1 - 2 | 250 | 92 | |
| GenBank mRNA | AK121401.1 | - | 1 - 2 | 138 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771656.1 transcripts.
Set Membership: |
|

LOC4328083
Genomic Map