Assay Details
| Gene Symbol: LOC4329131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772105.2 | XP_015627591.1 | 3 - 4 | 575 | 65 | |
| GenBank mRNA | AK061035.1 | - | 3 - 4 | 564 | 65 | |
More Information
Set Membership: |
|

LOC4329131
Genomic Map