Assay Details
| Gene Symbol: LOC4329835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772160.2 | XP_015627646.1 | 5 - 6 | 2235 | 96 | |
| XM_015772161.2 | XP_015627647.1 | 5 - 6 | 2235 | 96 | ||
| GenBank mRNA | AK102601.1 | - | 5 - 6 | 2212 | 96 | |
More Information
Set Membership: |
|

LOC4329835
Genomic Map