Assay Details
| Gene Symbol: LOC4329835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772160.2 | XP_015627646.1 | 8 - 9 | 2444 | 100 | |
| GenBank mRNA | AK102601.1 | - | 8 - 9 | 2421 | 100 | |
More Information
Set Membership: |
|

LOC4329835
Genomic Map