Assay Details
| Gene Symbol: LOC4331585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775485.2 | XP_015630971.1 | 1 - 2 | 2631 | 71 | |
| GenBank mRNA | AK121451.1 | - | 1 - 2 | 2633 | 71 | |
More Information
Set Membership: |
|

LOC4331585
Genomic Map