Assay Details
| Gene Symbol: LOC4331585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775485.2 | XP_015630971.1 | 3 - 4 | 3035 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4331585| Gene Symbol: LOC4331585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775485.2 | XP_015630971.1 | 3 - 4 | 3035 | 100 | |
Set Membership: |
|