Assay Details
| Gene Symbol: LOC4331585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775485.2 | XP_015630971.1 | 4 - 5 | 3716 | 91 | |
| GenBank mRNA | AK064325.1 | - | 1 - 2 | 134 | 91 | |
| AK121451.1 | - | 4 - 5 | 3718 | 91 | ||
More Information
Set Membership: |
|

LOC4331585
Genomic Map