Assay Details
| Gene Symbol: LOC4331585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775485.2 | XP_015630971.1 | 5 - 6 | 3878 | 124 | |
| GenBank mRNA | AK064325.1 | - | 2 - 3 | 296 | 124 | |
| AK121451.1 | - | 5 - 6 | 3880 | 124 | ||
More Information
Set Membership: |
|

LOC4331585
Genomic Map