Assay Details
| Gene Symbol: LOC4331585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775485.2 | XP_015630971.1 | 7 - 8 | 4387 | 72 | |
| GenBank mRNA | AK064325.1 | - | 4 - 5 | 805 | 72 | |
More Information
Set Membership: |
|

LOC4331585
Genomic Map