Assay Details
| Gene Symbol: LOC4331717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777127.2 | XP_015632613.1 | 17 - 18 | 2103 | 119 | |
| GenBank mRNA | AK065571.1 | - | 17 - 18 | 2031 | 119 | |
More Information
Set Membership: |
|

LOC4331717
Genomic Map