Assay Details
| Gene Symbol: LOC4331824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775389.2 | XP_015630875.1 | 5 - 6 | 1631 | 73 | |
| GenBank mRNA | AK121637.1 | - | 5 - 6 | 1509 | 73 | |
More Information
Set Membership: |
|

LOC4331824
Genomic Map