Assay Details
| Gene Symbol: LOC4331824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775389.2 | XP_015630875.1 | 8 - 9 | 2306 | 97 | |
| GenBank mRNA | AK109440.2 | - | 2 - 3 | 349 | 97 | |
| AK121637.1 | - | 8 - 9 | 2184 | 97 | ||
More Information
Set Membership: |
|

LOC4331824
Genomic Map