Assay Details
| Gene Symbol: LOC4331854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777612.2 | XP_015633098.1 | 10 - 11 | 1445 | 85 | |
| XM_015777614.2 | XP_015633100.1 | 10 - 11 | 1518 | 85 | ||
| XM_015777615.2 | XP_015633101.1 | 10 - 11 | 1426 | 85 | ||
More Information
Set Membership: |
|

LOC4331854
Genomic Map