Assay Details
| Gene Symbol: LOC4332578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773015.2 | XP_015628501.1 | 13 - 14 | 1876 | 76 | |
| GenBank mRNA | AK106465.1 | - | 1 - 2 | 131 | 76 | |
More Information
Set Membership: |
|

LOC4332578
Genomic Map