Assay Details
| Gene Symbol: LOC4332578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773015.2 | XP_015628501.1 | 15 - 16 | 2295 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4332578| Gene Symbol: LOC4332578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773015.2 | XP_015628501.1 | 15 - 16 | 2295 | 150 | |
Set Membership: |
|