Assay Details
| Gene Symbol: LOC4333818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777283.2 | XP_015632769.1 | 1 - 2 | 270 | 79 | |
| GenBank mRNA | AB099850.1 | - | 1 - 2 | 249 | 79 | |
| AK103565.1 | - | 1 - 2 | 257 | 79 | ||
| EU032588.1 | - | 1 - 2 | 183 | 79 | ||
More Information
Set Membership: |
|

LOC4333818
Genomic Map