Assay Details
| Gene Symbol: LOC4333818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777283.2 | XP_015632769.1 | 2 - 3 | 568 | 81 | |
| GenBank mRNA | AK103565.1 | - | 2 - 3 | 555 | 81 | |
More Information
Set Membership: |
|

LOC4333818
Genomic Map