Assay Details
| Gene Symbol: LOC4333818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777283.2 | XP_015632769.1 | 5 - 6 | 1221 | 80 | |
| GenBank mRNA | AB099850.1 | - | 5 - 6 | 1200 | 80 | |
| AK103565.1 | - | 5 - 6 | 1208 | 80 | ||
| EU032588.1 | - | 6 - 7 | 1146 | 80 | ||
| FJ660621.1 | - | 1 - 2 | 82 | 80 | ||
More Information
Set Membership: |
|

LOC4333818
Genomic Map