Assay Details
| Gene Symbol: LOC107276918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776040.2 | XP_015631526.1 | 1 | 160 | 128 | |
| GenBank mRNA | AK287828.1 | - | 1 | 150 | 128 | |
More Information
Set Membership: |
|

LOC107276918
Genomic Map