Assay Details
| Gene Symbol: LOC4337172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781193.1 | XP_015636679.1 | 1 - 2 | 1389 | 103 | |
| XM_015781194.2 | XP_015636680.1 | 2 - 3 | 495 | 103 | ||
| XM_015781195.2 | XP_015636681.1 | 2 - 3 | 489 | 103 | ||
| GenBank mRNA | AK062620.1 | - | 2 - 3 | 231 | 103 | |
| AK109401.2 | - | 2 - 3 | 227 | 103 | ||
More Information
Set Membership: |
|

LOC4337172
Genomic Map