Assay Details
| Gene Symbol: LOC4337276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779839.2 | XP_015635325.1 | 3 - 4 | 540 | 110 | |
| GenBank mRNA | AK059302.1 | - | 3 - 4 | 396 | 110 | |
| AK072534.1 | - | 3 - 4 | 486 | 110 | ||
| AK122053.1 | - | 431 | 110 | |||
More Information
Set Membership: |
|

LOC4337276
Genomic Map