Assay Details
| Gene Symbol: LOC4337276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779839.2 | XP_015635325.1 | 4 - 5 | 694 | 93 | |
| GenBank mRNA | AK059302.1 | - | 4 - 5 | 550 | 93 | |
| AK072534.1 | - | 4 - 5 | 640 | 93 | ||
| AK122053.1 | - | 585 | 93 | |||
More Information
Set Membership: |
|

LOC4337276
Genomic Map