Assay Details
| Gene Symbol: LOC4337276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779839.2 | XP_015635325.1 | 5 - 6 | 791 | 63 | |
| GenBank mRNA | AK059302.1 | - | 5 - 6 | 647 | 63 | |
| AK072534.1 | - | 5 - 6 | 737 | 63 | ||
| AK122053.1 | - | 682 | 63 | |||
More Information
Set Membership: |
|

LOC4337276
Genomic Map