Assay Details
| Gene Symbol: LOC4338442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782427.2 | XP_015637913.1 | 2 - 3 | 513 | 139 | |
| GenBank mRNA | AK100884.1 | - | 2 - 3 | 386 | 139 | |
More Information
Set Membership: |
|

LOC4338442
Genomic Map