Assay Details
| Gene Symbol: LOC4340221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785722.2 | XP_015641208.2 | 5 - 6 | 695 | 61 | |
| GenBank mRNA | AK068888.1 | - | 5 - 6 | 521 | 61 | |
More Information
Set Membership: |
|

LOC4340221
Genomic Map