Assay Details
| Gene Symbol: LOC4340221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785722.2 | XP_015641208.2 | 7 - 8 | 840 | 63 | |
| GenBank mRNA | AK068888.1 | - | 7 - 8 | 666 | 63 | |
More Information
Set Membership: |
|

LOC4340221
Genomic Map