Assay Details
| Gene Symbol: LOC4341189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787602.2 | XP_015643088.1 | 5 - 6 | 1327 | 125 | |
| XM_015787603.1 | XP_015643089.1 | 4 - 5 | 1232 | 125 | ||
| GenBank mRNA | AK099993.1 | - | 5 - 6 | 1292 | 125 | |
More Information
Set Membership: |
|

LOC4341189
Genomic Map