Assay Details
| Gene Symbol: LOC4341358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788726.1 | XP_015644212.1 | 11 - 12 | 1745 | 77 | |
| GenBank mRNA | AK102756.1 | - | 11 - 12 | 1751 | 77 | |
| AK105619.1 | - | 2 - 3 | 427 | 77 | ||
More Information
Set Membership: |
|

LOC4341358
Genomic Map