Assay Details
| Gene Symbol: LOC4341358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788726.1 | XP_015644212.1 | 12 - 13 | 1852 | 58 | |
| GenBank mRNA | AK102756.1 | - | 12 - 13 | 1858 | 58 | |
| AK105619.1 | - | 3 - 4 | 534 | 58 | ||
More Information
Set Membership: |
|

LOC4341358
Genomic Map