Assay Details
| Gene Symbol: LOC4341358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788726.1 | XP_015644212.1 | 4 - 5 | 773 | 67 | |
| GenBank mRNA | AK102756.1 | - | 4 - 5 | 779 | 67 | |
More Information
Set Membership: |
|

LOC4341358
Genomic Map