Assay Details
| Gene Symbol: LOC4341457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786499.2 | XP_015641985.1 | 2 - 3 | 720 | 64 | |
| XM_015786500.2 | XP_015641986.1 | 2 - 3 | 720 | 64 | ||
| GenBank mRNA | AK106474.1 | - | 2 - 3 | 282 | 64 | |
More Information
Set Membership: |
|

LOC4341457
Genomic Map