Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 9 - 10 | 2100 | 70 | |
| GenBank mRNA | AB096015.1 | - | 9 - 10 | 1963 | 70 | |
| AB120430.1 | - | 9 - 10 | 2106 | 70 | ||
| AK058530.1 | - | 2 - 3 | 468 | 70 | ||
| AK069977.1 | - | 10 - 11 | 1740 | 70 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map