Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 12 - 13 | 2298 | 118 | |
| GenBank mRNA | AB096015.1 | - | 12 - 13 | 2161 | 118 | |
| AB120430.1 | - | 12 - 13 | 2304 | 118 | ||
| AK058530.1 | - | 5 - 6 | 666 | 118 | ||
| AK069977.1 | - | 13 - 14 | 1938 | 118 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map