Assay Details
Gene Symbol: LOC4341695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788508.2 | XP_015643994.1 | 15 - 16 | 2623 | 74 | |
GenBank mRNA | AB096015.1 | - | 15 - 16 | 2486 | 74 | |
AB120430.1 | - | 15 - 16 | 2629 | 74 | ||
AK058530.1 | - | 8 - 9 | 991 | 74 | ||
AK069977.1 | - | 16 - 17 | 2263 | 74 | ||
U34283.1 | - | 3 - 4 | 305 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
