Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 17 - 18 | 2782 | 84 | |
| GenBank mRNA | AB096015.1 | - | 17 - 18 | 2645 | 84 | |
| AB120430.1 | - | 17 - 18 | 2788 | 84 | ||
| AK058530.1 | - | 10 - 11 | 1150 | 84 | ||
| AK069977.1 | - | 18 - 19 | 2422 | 84 | ||
| U34283.1 | - | 5 - 6 | 464 | 84 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map