Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 18 - 19 | 2891 | 64 | |
| GenBank mRNA | AB096015.1 | - | 18 - 19 | 2754 | 64 | |
| AB120430.1 | - | 18 - 19 | 2897 | 64 | ||
| AK058530.1 | - | 11 - 12 | 1259 | 64 | ||
| AK069977.1 | - | 19 - 20 | 2531 | 64 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map