Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 6 - 7 | 1439 | 112 | |
| GenBank mRNA | AB096015.1 | - | 6 - 7 | 1302 | 112 | |
| AB120430.1 | - | 6 - 7 | 1445 | 112 | ||
| AK069977.1 | - | 7 - 8 | 1079 | 112 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map