Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 8 - 9 | 1682 | 95 | |
| GenBank mRNA | AB096015.1 | - | 8 - 9 | 1545 | 95 | |
| AB120430.1 | - | 8 - 9 | 1688 | 95 | ||
| AK058530.1 | - | 1 - 2 | 50 | 95 | ||
| AK069977.1 | - | 9 - 10 | 1322 | 95 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map