Assay Details
| Gene Symbol: LOC4342073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785700.2 | XP_015641186.1 | 7 - 8 | 857 | 118 | |
| GenBank mRNA | AK059304.1 | - | 7 - 8 | 812 | 118 | |
| AK102724.1 | - | 7 - 8 | 858 | 118 | ||
More Information
Set Membership: |
|

LOC4342073
Genomic Map