Assay Details
| Gene Symbol: LOC4344937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794913.2 | XP_015650399.1 | 1 - 2 | 759 | 71 | |
| GenBank mRNA | AB126089.1 | - | 1 - 2 | 731 | 71 | |
| AK105258.1 | - | 1 - 2 | 765 | 71 | ||
More Information
Set Membership: |
|

LOC4344937
Genomic Map