Assay Details
| Gene Symbol: LOC9269435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793020.2 | XP_015648506.1 | 16 - 17 | 4023 | 67 | |
| GenBank mRNA | AK069036.1 | - | 14 - 15 | 2699 | 67 | |
| AK100610.1 | - | 13 - 14 | 2371 | 67 | ||
More Information
Set Membership: |
|

LOC9269435
Genomic Map