Assay Details
| Gene Symbol: LOC9269435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793020.2 | XP_015648506.1 | 18 - 19 | 4161 | 70 | |
| GenBank mRNA | AK069036.1 | - | 16 - 17 | 2837 | 70 | |
| AK100610.1 | - | 15 - 16 | 2509 | 70 | ||
More Information
Set Membership: |
|

LOC9269435
Genomic Map