Assay Details
| Gene Symbol: LOC9269435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793020.2 | XP_015648506.1 | 19 - 20 | 4570 | 65 | |
| GenBank mRNA | AK069036.1 | - | 17 - 18 | 3246 | 65 | |
More Information
Set Membership: |
|

LOC9269435
Genomic Map