Assay Details
| Gene Symbol: LOC9269435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793020.2 | XP_015648506.1 | 2 - 3 | 1084 | 117 | |
| XM_015793025.2 | XP_015648511.1 | 2 - 3 | 1084 | 117 | ||
| GenBank mRNA | AK069036.1 | - | 2 - 3 | 1027 | 117 | |
More Information
Set Membership: |
|

LOC9269435
Genomic Map