Assay Details
| Gene Symbol: LOC9269435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793020.2 | XP_015648506.1 | 6 - 7 | 3187 | 88 | |
| XM_015793025.2 | XP_015648511.1 | 6 - 7 | 3187 | 88 | ||
| GenBank mRNA | AK069036.1 | - | 4 - 5 | 1863 | 88 | |
| AK100610.1 | - | 3 - 4 | 1535 | 88 | ||
More Information
Set Membership: |
|

LOC9269435
Genomic Map