Assay Details
| Gene Symbol: LOC9269435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793020.2 | XP_015648506.1 | 11 - 12 | 3603 | 85 | |
| XM_015793025.2 | XP_015648511.1 | 11 - 12 | 3603 | 85 | ||
| GenBank mRNA | AK069036.1 | - | 9 - 10 | 2279 | 85 | |
| AK100610.1 | - | 8 - 9 | 1951 | 85 | ||
More Information
Set Membership: |
|

LOC9269435
Genomic Map