Assay Details
| Gene Symbol: LOC4345288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795587.2 | XP_015651073.1 | 2 - 3 | 603 | 114 | |
| XM_015795588.2 | XP_015651074.1 | 2 - 3 | 603 | 114 | ||
| GenBank mRNA | AK102227.1 | - | 2 - 3 | 604 | 114 | |
| AK105392.1 | - | 2 - 3 | 561 | 114 | ||
More Information
Set Membership: |
|

LOC4345288
Genomic Map