Assay Details
| Gene Symbol: LOC4345288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795587.2 | XP_015651073.1 | 6 - 7 | 1249 | 86 | |
| XM_015795588.2 | XP_015651074.1 | 6 - 7 | 1249 | 86 | ||
| GenBank mRNA | AK105392.1 | - | 6 - 7 | 1207 | 86 | |
More Information
Set Membership: |
|

LOC4345288
Genomic Map