Assay Details
| Gene Symbol: LOC4345652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794833.2 | XP_015650319.1 | 1 - 2 | 338 | 66 | |
| GenBank mRNA | AJ495796.1 | - | 1 - 2 | 312 | 66 | |
| AK059468.1 | - | 1 - 2 | 210 | 66 | ||
More Information
Set Membership: |
|

LOC4345652
Genomic Map