Assay Details
| Gene Symbol: LOC4345652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794833.2 | XP_015650319.1 | 2 - 3 | 464 | 67 | |
| GenBank mRNA | AJ495796.1 | - | 2 - 3 | 438 | 67 | |
More Information
Set Membership: |
|

LOC4345652
Genomic Map